Gene Transcripts

Gene Symbol
NLRP13
Entrez Gene ID
126204
Gene Name
NLR family pyrin domain containing 13
Gene Aliases
CLR19.7, NALP13, NOD14, PAN13
Chromosome Location
Chr. 19: 55891692 - 55932336 on Build GRCh38
UniGene ID
Hs.446924
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321057.1 NP_001307986.1 11 3080
NM_176810.2 NP_789780.2 11 3080
GenBank mRNA AY154468.1 - 11 3055

View Full Product Details