Gene Transcripts

Gene Symbol
OR10H4
Entrez Gene ID
126541
Gene Name
olfactory receptor family 10 subfamily H member 4
Gene Aliases
OR19-28
Chromosome Location
Chr. 19: 15949008 - 15949958 on Build GRCh38
UniGene ID
Hs.553579
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004465.1 NP_001004465.1 1 479

View Full Product Details