Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001139456.1 | NP_001132928.1 | 9 | 818 | |
NM_174959.3 | NP_777619.1 | 5 | 435 | ||
XM_005250143.3 | XP_005250200.1 | 10 | 1222 | ||
XM_011515797.2 | XP_011514099.1 | 6 | 509 | ||
XM_017011746.1 | XP_016867235.1 | 7 | 624 | ||
XM_017011747.1 | XP_016867236.1 | 6 | 449 | ||
XM_017011748.1 | XP_016867237.1 | 6 | 454 | ||
GenBank mRNA | BC036796.1 | - | 5 | 435 | |
BX648789.1 | - | 5 | 4055 |