Gene Transcripts
- Gene Symbol
- RALYL
- Entrez Gene ID
- 138046
- Gene Name
- RALY RNA binding protein-like
- Gene Aliases
- HNRPCL3
- Chromosome Location
- Chr. 8: 84182629 - 84921844 on Build GRCh38
- UniGene ID
- Hs.121663
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001100391.1 |
NP_001093861.1
|
5, 6 |
576 |
|
NM_001100392.1 |
NP_001093862.1
|
6, 7 |
819 |
|
NM_001100393.1 |
NP_001093863.1
|
5, 6 |
787 |
|
NM_001287243.1 |
NP_001274172.1
|
4, 5 |
1467 |
|
NM_001287244.1 |
NP_001274173.1
|
4, 5 |
281 |
|
NM_173848.5 |
NP_776247.3
|
5, 6 |
1500 |
|
XM_011517462.2 |
XP_011515764.1
|
5, 6 |
1571 |
|
XM_017013058.1 |
XP_016868547.1
|
4, 5 |
742 |
|
XM_017013059.1 |
XP_016868548.1
|
7, 8 |
870 |
|
XM_017013060.1 |
XP_016868549.1
|
5, 6 |
3020 |
|
XM_017013061.1 |
XP_016868550.1
|
5, 6 |
2172 |
|
XM_017013062.1 |
XP_016868551.1
|
7, 8 |
979 |
|
XM_017013063.1 |
XP_016868552.1
|
5, 6 |
742 |
|
XM_017013064.1 |
XP_016868553.1
|
6, 7 |
1093 |
|
XM_017013065.1 |
XP_016868554.1
|
5, 6 |
641 |
|
XM_017013066.1 |
XP_016868555.1
|
4, 5 |
754 |
|
XM_017013067.1 |
XP_016868556.1
|
5, 6 |
1645 |
|
XM_017013068.1 |
XP_016868557.1
|
6, 7 |
837 |
|
XM_017013069.1 |
XP_016868558.1
|
6, 7 |
893 |
|
XM_017013070.1 |
XP_016868559.1
|
5, 6 |
737 |
|
XM_017013071.1 |
XP_016868560.1
|
4, 5 |
608 |
|
XM_017013072.1 |
XP_016868561.1
|
5, 6 |
728 |
|
XM_017013073.1 |
XP_016868562.1
|
5, 6 |
728 |
|
XM_017013074.1 |
XP_016868563.1
|
5, 6 |
641 |
|
XM_017013075.1 |
XP_016868564.1
|
4, 5 |
693 |
|
XM_017013076.1 |
XP_016868565.1
|
5, 6 |
521 |
|
XM_017013077.1 |
XP_016868566.1
|
6, 7 |
1771 |
|
XM_017013078.1 |
XP_016868567.1
|
5, 6 |
787 |
|
XM_017013079.1 |
XP_016868568.1
|
6, 7 |
770 |
|
XM_017013080.1 |
XP_016868569.1
|
4, 5 |
488 |
|
XM_017013081.1 |
XP_016868570.1
|
4, 5 |
488 |
|
XM_017013082.1 |
XP_016868571.1
|
4, 5 |
754 |
|
XM_017013083.1 |
XP_016868572.1
|
4, 5 |
442 |
|
XM_017013084.1 |
XP_016868573.1
|
3, 4 |
1277 |
|
XM_017013085.1 |
XP_016868574.1
|
5, 6 |
324 |
|
XM_017013086.1 |
XP_016868575.1
|
4, 5 |
281 |
|
XM_017013087.1 |
XP_016868576.1
|
4, 5 |
281 |
|
GenBank mRNA |
AF495529.1 |
- |
5, 6 |
1500 |
|
AF495530.1 |
- |
5, 6 |
717 |
|
AK094543.1 |
- |
5, 6 |
731 |
|
AK095026.1 |
- |
4, 5 |
1283 |
|
AK095560.1 |
- |
6, 7 |
770 |
|
AK123437.1 |
- |
4, 5 |
281 |
|
BC031090.1 |
- |
5, 6 |
565 |
5311935 |
View Full Product Details