Gene Transcripts

Gene Symbol
ZNF569
Entrez Gene ID
148266
Gene Name
zinc finger protein 569
Gene Aliases
ZAP1, ZNF
Chromosome Location
Chr. 19: 37411146 - 37469268 on Build GRCh38
UniGene ID
Hs.511848
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152484.2 NP_689697.2 6 856
XM_006723046.2 XP_006723109.1 8 1183
XM_006723047.3 XP_006723110.1 7 1106
XM_006723048.3 XP_006723111.1 7 1030
XM_011526538.2 XP_011524840.1 7 945
XM_011526539.2 XP_011524841.1 5 439
XM_017026376.1 XP_016881865.1 6 691
XM_017026377.1 XP_016881866.1 6 770
XM_017026378.1 XP_016881867.1 7 808
XM_017026379.1 XP_016881868.1 4 657
XM_017026380.1 XP_016881869.1 4 465
XM_017026381.1 XP_016881870.1 5 868
XM_017026382.1 XP_016881871.1 4 565
GenBank mRNA AK122846.1 - 3 177
AK289987.1 - 6 629
AL833408.1 - 6 857
AY835704.1 - 6 856
BC059407.1 - 5 484
BC117219.1 - 2 113 40125770

View Full Product Details