Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001015887.1 | NP_001015887.1 | 7 | 1474 | |
NM_152538.2 | NP_689751.2 | 9 | 1605 | ||
XM_006713517.1 | XP_006713580.1 | 8 | 1525 | ||
XM_011512462.2 | XP_011510764.1 | 8 | 1683 | ||
XM_011512463.1 | XP_011510765.1 | 9 | 1428 | ||
XM_011512464.2 | XP_011510766.1 | 9 | 1416 | ||
XM_011512465.1 | XP_011510767.1 | 9 | 1470 | ||
XM_011512466.2 | XP_011510768.1 | 8 | 1615 | ||
XM_011512467.2 | XP_011510769.1 | 8 | 1602 | ||
XM_011512468.1 | XP_011510770.1 | 10 | 1618 | ||
XM_011512469.1 | XP_011510771.1 | 8 | 1278 | ||
XM_011512470.1 | XP_011510772.1 | 8 | 1493 | ||
XM_011512471.2 | XP_011510773.1 | 8 | 1531 | ||
XM_011512472.1 | XP_011510774.1 | 8 | 1294 | ||
XM_017005789.1 | XP_016861278.1 | 7 | 1263 | ||
XM_017005790.1 | XP_016861279.1 | 8 | 1453 | ||
GenBank mRNA | AB071618.1 | - | 7 | 1464 | |
AB079879.1 | - | 7 | 1227 | ||
AK097667.1 | - | 9 | 1517 | ||
AK122611.1 | - | 7 | 1365 | ||
AK292387.1 | - | 8 | 1493 | ||
AK296503.1 | - | 7 | 1289 | ||
AL834256.1 | - | 5 | 905 | ||
AY358141.1 | - | 7 | 1515 | ||
BC034411.1 | - | 9 | 1398 | 5172176 |