Gene Transcripts
- Gene Symbol
- IGSF11
- Entrez Gene ID
- 152404
- Gene Name
- immunoglobulin superfamily member 11
- Gene Aliases
- BT-IgSF, CT119, CXADRL1, Igsf13, VSIG3
- Chromosome Location
- Chr. 3: 118900630 - 119146068 on Build GRCh38
- UniGene ID
- Hs.112873
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001015887.1 |
NP_001015887.1
|
5, 6 |
961 |
|
NM_152538.2 |
NP_689751.2
|
7, 8 |
1092 |
|
XM_006713517.1 |
XP_006713580.1
|
6, 7 |
1012 |
|
XM_011512462.2 |
XP_011510764.1
|
6, 7 |
1170 |
|
XM_011512463.1 |
XP_011510765.1
|
7, 8 |
915 |
|
XM_011512464.2 |
XP_011510766.1
|
7, 8 |
903 |
|
XM_011512465.1 |
XP_011510767.1
|
7, 8 |
957 |
|
XM_011512466.2 |
XP_011510768.1
|
6, 7 |
1102 |
|
XM_011512468.1 |
XP_011510770.1
|
8, 9 |
1105 |
|
XM_011512469.1 |
XP_011510771.1
|
6, 7 |
765 |
|
XM_011512470.1 |
XP_011510772.1
|
6, 7 |
980 |
|
XM_011512472.1 |
XP_011510774.1
|
6, 7 |
781 |
|
XM_017005789.1 |
XP_016861278.1
|
5, 6 |
750 |
|
XM_017005790.1 |
XP_016861279.1
|
6, 7 |
940 |
|
GenBank mRNA |
AB071618.1 |
- |
5, 6 |
951 |
|
AB079879.1 |
- |
5, 6 |
714 |
|
AK097667.1 |
- |
7, 8 |
1004 |
|
AK122611.1 |
- |
5, 6 |
852 |
|
AK292387.1 |
- |
6, 7 |
980 |
|
AL834256.1 |
- |
3, 4 |
392 |
|
AY358141.1 |
- |
5, 6 |
1002 |
|
BC034411.1 |
- |
7, 8 |
885 |
5172176 |
View Full Product Details