Gene Transcripts

Gene Symbol
PEBP4
Entrez Gene ID
157310
Gene Name
phosphatidylethanolamine binding protein 4
Gene Aliases
CORK-1, CORK1, GWTM1933, HEL-S-300, PEBP-4, PRO4408, hPEBP4
Chromosome Location
Chr. 8: 22713251 - 22941095 on Build GRCh38
UniGene ID
Hs.491242
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144962.2 NP_659399.2 6 543
XM_011544414.2 XP_011542716.1 6 598
GenBank mRNA AY037148.1 - 6 516
AY359109.1 - 6 539
AY730275.1 - 6 494
BC020779.1 - 6 535 4700840
BF853000.1 - 6 543
FJ224332.1 - 6 535

View Full Product Details