Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_178422.5 | NP_848509.1 | 2 | 1226 | |
XM_005245745.3 | XP_005245802.1 | 3 | 1048 | ||
XM_005245746.3 | XP_005245803.1 | 2 | 964 | ||
XM_011540861.1 | XP_011539163.1 | 3 | 871 | ||
XM_011540862.2 | XP_011539164.1 | 4 | 1137 | ||
GenBank mRNA | AF313620.1 | - | 2 | 655 | |
AY424285.1 | - | 1 | 787 | ||
BC034015.1 | - | 2 | 1226 | 4400504 | |
BC127800.1 | - | 1 | 337 |