Gene Transcripts
- Gene Symbol
- SLC30A8
- Entrez Gene ID
- 169026
- Gene Name
- solute carrier family 30 member 8
- Gene Aliases
- ZNT8, ZnT-8
- Chromosome Location
- Chr. 8: 116950273 - 117176714 on Build GRCh38
- UniGene ID
- Hs.532270
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001172811.1 |
NP_001166282.1
|
9, 10 |
1125 |
|
NM_001172813.1 |
NP_001166284.1
|
10, 11 |
1370 |
|
NM_001172814.1 |
NP_001166285.1
|
8, 9 |
1212 |
|
NM_001172815.2 |
NP_001166286.1
|
10, 11 |
1239 |
|
NM_173851.2 |
NP_776250.2
|
7, 8 |
1182 |
|
GenBank mRNA |
AK298294.1 |
- |
11, 12 |
1373 |
|
AL713790.1 |
- |
10, 11 |
1211 |
|
AY117411.1 |
- |
7, 8 |
952 |
|
BC126446.1 |
- |
8, 9 |
1051 |
|
BC143953.1 |
- |
8, 9 |
1052 |
|
EF560713.1 |
- |
9, 10 |
1125 |
|
View Full Product Details