Gene Transcripts
- Gene Symbol
- RBM24
- Entrez Gene ID
- 221662
- Gene Name
- RNA binding motif protein 24
- Gene Aliases
- RNPC6, dJ259A10.1
- Chromosome Location
- Chr. 6: 17281508 - 17293875 on Build GRCh38
- UniGene ID
- Hs.519904
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001143941.1 |
NP_001137413.1
|
3, 4 |
324 |
|
NM_001143942.1 |
NP_001137414.1
|
3, 4 |
340 |
|
NM_153020.2 |
NP_694565.1
|
2, 3 |
272 |
|
XM_011514388.2 |
XP_011512690.1
|
1, 2 |
24 |
|
XM_011514389.2 |
XP_011512691.1
|
1, 2 |
18 |
|
GenBank mRNA |
AK055391.1 |
- |
2, 3 |
272 |
|
AK095016.1 |
- |
3, 4 |
375 |
|
AK293052.1 |
- |
3, 4 |
340 |
|
AK315856.1 |
- |
3, 4 |
340 |
|
AY547318.1 |
- |
3, 4 |
336 |
|
BC104808.1 |
- |
2, 3 |
240 |
8143811 |
BC104810.1 |
- |
2, 3 |
240 |
8143813 |
BP309176.1 |
- |
3, 4 |
324 |
|
View Full Product Details