Gene Transcripts

Gene Symbol
FFAR4
Entrez Gene ID
338557
Gene Name
free fatty acid receptor 4
Gene Aliases
BMIQ10, GPR120, GPR129, GT01, O3FAR1, PGR4
Chromosome Location
Chr. 10: 93566179 - 93590072 on Build GRCh38
UniGene ID
Hs.661022
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195755.1 NP_001182684.1 3 1128
NM_181745.3 NP_859529.2 4 1176
GenBank mRNA AB115768.1 - 4 1120
AK301525.1 - 3 1101
AY288417.1 - 4 1117
BC101175.2 - 3 1072 40016354

View Full Product Details