Gene Transcripts

Gene Symbol
TOMM20L
Entrez Gene ID
387990
Gene Name
translocase of outer mitochondrial membrane 20 like
Gene Aliases
UNQ9438
Chromosome Location
Chr. 14: 58395904 - 58417080 on Build GRCh38
UniGene ID
Hs.592307
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207377.2 NP_997260.1 2, 3 219
GenBank mRNA AY358266.1 - 2, 3 231
BC137557.1 - 2, 3 217 9021557
BC137563.1 - 2, 3 217 9021563

View Full Product Details