Gene Transcripts

Gene Symbol
OR6C70
Entrez Gene ID
390327
Gene Name
olfactory receptor family 6 subfamily C member 70
Gene Aliases
Chromosome Location
Chr. 12: 55469200 - 55470138 on Build GRCh38
UniGene ID
Hs.632718
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005499.1 NP_001005499.1 1 430

View Full Product Details