Gene Transcripts

Gene Symbol
OR6C65
Entrez Gene ID
403282
Gene Name
olfactory receptor family 6 subfamily C member 65
Gene Aliases
Chromosome Location
Chr. 12: 55400529 - 55401467 on Build GRCh38
UniGene ID
Hs.554553
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005518.1 NP_001005518.1 1 218
GenBank mRNA BC136897.1 - 1 317
BC136899.1 - 1 317 9020890

View Full Product Details