Gene Transcripts

Gene Symbol
KCNE1
Entrez Gene ID
3753
Gene Name
potassium voltage-gated channel subfamily E regulatory subunit 1
Gene Aliases
ISK, JLNS, JLNS2, LQT2/5, LQT5, MinK
Chromosome Location
Chr. 21: 34446688 - 34512275 on Build GRCh38
UniGene ID
Hs.121495
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000219.5 NP_000210.2 4 769
NM_001127668.3 NP_001121140.1 3 381
NM_001127669.3 NP_001121141.1 3 353
NM_001127670.3 NP_001121142.1 2 618
NM_001270402.2 NP_001257331.1 3 544
NM_001270403.2 NP_001257332.1 3 492
NM_001270404.2 NP_001257333.1 2 433
NM_001270405.2 NP_001257334.1 3 345
XM_017028342.1 XP_016883831.1 2 2121
GenBank mRNA AF135188.1 - 1 144
AK304336.1 - 3 437
AK313543.1 - 3 376
AY789479.1 - 3 308
AY789480.1 - 2 550
BC036452.1 - 3 355 5266786
BC046224.1 - 4 769 6198685
CR541756.1 - 1 144
DA806205.1 - 3 345
DB214219.1 - 2 393
DB223354.1 - 2 424
DB230009.1 - 3 467
DB233067.1 - 3 534
EF514882.1 - 1 144
EF514883.1 - 1 144
L28168.1 - 1 162

Gene Symbol
KCNE1B
Entrez Gene ID
102723475
Gene Name
potassium voltage-gated channel subfamily E regulatory subunit 1B
Gene Aliases
Chromosome Location
Chr. 21: 7816677 - 7829632 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006723946.2 XP_006724009.1 2 574
XM_006723949.2 XP_006724012.1 3 421

View Full Product Details