Gene Transcripts

Gene Symbol
NDUFB8
Entrez Gene ID
4714
Gene Name
NADH:ubiquinone oxidoreductase subunit B8
Gene Aliases
ASHI, CI-ASHI
Chromosome Location
Chr. 10: 100523729 - 100529923 on Build GRCh38
UniGene ID
Hs.523215
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001284367.1 NP_001271296.1 5 799
NM_001284368.1 NP_001271297.1 5 498
NM_005004.3 NP_004995.1 5 589
GenBank mRNA AA782554.1 - 1 166
AF044958.1 - 5 539
AF115968.1 - 5 545
AI282606.1 - 1 139
AK289579.1 - 5 530
AL080056.1 - 5 549
BC000466.2 - 5 530 2822781
BC019276.1 - 5 530 2822781
BM008944.1 - 5 486
CR533490.1 - 5 517

View Full Product Details