Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002922.3 | NP_002913.3 | 5 | 680 | |
GenBank mRNA | AF493925.1 | - | 5 | 575 | |
AK093544.1 | - | 4 | 2018 | ||
AK315607.1 | - | 5 | 634 | ||
BC015510.1 | - | 5 | 612 | 3916789 | |
BT006668.1 | - | 5 | 575 | ||
EU668340.1 | - | 5 | 612 | ||
X73427.1 | - | 5 | 589 |