Gene Transcripts

Gene Symbol
DYNLT1
Entrez Gene ID
6993
Gene Name
dynein light chain Tctex-type 1
Gene Aliases
CW-1, TCTEL1, tctex-1
Chromosome Location
Chr. 6: 158636474 - 158644786 on Build GRCh38
UniGene ID
Hs.445999
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291602.1 NP_001278531.1 4 363
NM_001291603.1 NP_001278532.1 5 385
NM_006519.3 NP_006510.1 5 405
GenBank mRNA AK026669.1 - 2 2529
AK315601.1 - 5 362
BC029412.1 - 5 358 4074224
BC105588.1 - 5 354 6723966
BU675683.1 - 1 380
D50663.1 - 5 339
DN915418.1 - 4 299
EU862237.1 - 5 331
HY017024.1 - 4 316
HY171700.1 - 5 408
U56255.1 - 5 327

View Full Product Details