Gene Transcripts

Gene Symbol
ARL4C
Entrez Gene ID
10123
Gene Name
ADP ribosylation factor like GTPase 4C
Gene Aliases
ARL7, LAK
Chromosome Location
Chr. 2: 234493041 - 234497053 on Build GRCh38
UniGene ID
Hs.111554
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282431.1 NP_001269360.1 1 652
NM_005737.3 NP_005728.2 1 648
GenBank mRNA AB016811.1 - 1 156
AF493892.1 - 1 185
AJ579850.1 - 1 199
AJ579851.1 - 1 199
AK314119.1 - 1 189
BC089043.1 - 1 652 6578402
BT019378.1 - 1 185
BT019379.1 - 1 185
Y17804.1 - 1 199

View Full Product Details