Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006319.4 | NP_006310.1 | 2 | 490 | |
GenBank mRNA | AF014807.1 | - | 2 | 406 | |
AK094468.1 | - | 1 | 324 | ||
AK097691.1 | - | 2 | 936 | ||
AK131349.1 | - | 2 | 396 | ||
AK222676.1 | - | 2 | 369 | ||
AK290632.1 | - | 3 | 264 | ||
AK310988.1 | - | 390 | |||
AK314544.1 | - | 2 | 421 | ||
BC001444.2 | - | 2 | 326 | 3139134 | |
BI490060.1 | - | 1 | 318 | ||
BT007301.1 | - | 2 | 59 | ||
CR542016.1 | - | 2 | 59 |