Gene Transcripts

Gene Symbol
MORF4L1
Entrez Gene ID
10933
Gene Name
mortality factor 4 like 1
Gene Aliases
Eaf3, FWP006, HsT17725, MEAF3, MORFRG15, MRG15, S863-6
Chromosome Location
Chr. 15: 78872781 - 78897739 on Build GRCh38
UniGene ID
Hs.374503
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001265603.1 NP_001252532.1 2 299
NM_001265604.1 NP_001252533.1 4 367
NM_001265605.1 NP_001252534.1 4 267
NM_006791.3 NP_006782.1 4 414
NM_206839.2 NP_996670.1 5 531
XM_017021872.1 XP_016877361.1 4 379
GenBank mRNA AF070664.1 - 4 221
AF100615.1 - 4 308
AF109188.1 - 4 203
AF161546.1 - 4 200
AF167173.1 - 4 216
AF218011.1 - 4 319
AK095823.1 - 5 816
AK098485.1 - 4 334
AK296650.1 - 5 422
AK300789.1 - 4 351
AK310181.1 - 3 152
AK310592.1 - 4 213
AK310702.1 - 4 222
AL137697.1 - 4 208
AY148481.1 - 5 425
BC002936.2 - 4 295 3955120
BC022845.2 - 4 295 2988803
BC067826.1 - 4 190 4824022
BC141840.1 - 4 222 8860303
BG771854.1 - 4 424
BX382045.2 - 4 211
CX165647.1 - 2 50
DB455429.1 - 4 267

View Full Product Details