Gene Transcripts

Gene Symbol
TNRC6B
Entrez Gene ID
23112
Gene Name
trinucleotide repeat containing 6B
Gene Aliases
Chromosome Location
Chr. 22: 40044817 - 40335808 on Build GRCh38
UniGene ID
Hs.372082
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024843.1 NP_001020014.1 18 2192
NM_001162501.1 NP_001155973.1 17 4513
NM_015088.2 NP_055903.2 15 4183
GenBank mRNA AB029016.2 - 15 4163
AK294519.1 - 17 4504
AK296590.1 - 8 1232
AK304087.1 - 12 1668
BC028626.2 - 18 2125 4826787
BX647630.1 - 12 3385

View Full Product Details