Gene Transcripts

Gene Symbol
SLC16A8
Entrez Gene ID
23539
Gene Name
solute carrier family 16 member 8
Gene Aliases
MCT3, REMP
Chromosome Location
Chr. 22: 38078137 - 38083163 on Build GRCh38
UniGene ID
Hs.745256
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013356.2 NP_037488.2 4 910
XM_017028685.1 XP_016884174.1 3 965
GenBank mRNA AF132610.1 - 4 889
BM714896.1 - 2 489

View Full Product Details