Gene Transcripts

Gene Symbol
SIGLEC7
Entrez Gene ID
27036
Gene Name
sialic acid binding Ig like lectin 7
Gene Aliases
AIRM1, CD328, CDw328, D-siglec, QA79, SIGLEC-7, SIGLEC19P, SIGLECP2, p75, p75/AIRM1
Chromosome Location
Chr. 19: 51142261 - 51153526 on Build GRCh38
UniGene ID
Hs.655393
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014385.3 NP_055200.1 6 1241
NM_016543.3 NP_057627.2 5 962
NR_102350.1 2 550
XM_011526721.2 XP_011525023.1 4 945
GenBank mRNA AF170485.1 - 6 1211
AF178981.1 - 5 948
AF193441.1 - 5 956
AJ007395.1 - 6 1236
AJ130710.1 - 6 1172
AJ130711.1 - 5 893
AJ130712.1 - 2 481
AK314875.1 - 6 1239
BC028150.1 - 6 1232 5217287

View Full Product Details