Gene Transcripts

Gene Symbol
ST6GALNAC4
Entrez Gene ID
27090
Gene Name
ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 4
Gene Aliases
IV, SIAT3-C, SIAT3C, SIAT7-D, SIAT7D, ST6GALNACIV, ST6GalNAc
Chromosome Location
Chr. 9: 127907886 - 127917052 on Build GRCh38
UniGene ID
Hs.3972
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175039.3 NP_778204.1 4 519
NM_175040.3 NP_778205.1 3 432
XM_011518527.2 XP_011516829.1 5 611
XM_011518529.2 XP_011516831.1 5 614
GenBank mRNA AB035172.1 - 3 264
AJ271734.1 - 4 534
AK000600.1 - 4 515
AK292049.1 - 3 423
BC036705.2 - 4 425 4309208
CB126299.1 - 3 432

View Full Product Details