Gene Transcripts

Gene Symbol
SERTAD3
Entrez Gene ID
29946
Gene Name
SERTA domain containing 3
Gene Aliases
RBT1
Chromosome Location
Chr. 19: 40440841 - 40444375 on Build GRCh38
UniGene ID
Hs.515412
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013368.3 NP_037500.2 2 585
NM_203344.2 NP_976219.1 2 474
XM_005258832.3 XP_005258889.1 3 682
XM_006723179.3 XP_006723242.1 2 534
GenBank mRNA AF192529.1 - 2 598
AK074604.1 - 2 399
BC014061.2 - 2 398 3926937
BC050643.2 - 2 474 6503515

View Full Product Details