Gene Transcripts
- Gene Symbol
- TERF2IP
- Entrez Gene ID
- 54386
- Gene Name
- TERF2 interacting protein
- Gene Aliases
- DRIP5, RAP1
- Chromosome Location
- Chr. 16: 75647704 - 75657442 on Build GRCh38
- UniGene ID
- Hs.301419
- Species
- Homo sapiens
- Gene Symbol
- LOC100288358
- Entrez Gene ID
- 100288358
- Gene Name
- uncharacterized LOC100288358
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.301419
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK298535.1 |
- |
1 |
743 |
|
View Full Product Details