Gene Transcripts

Gene Symbol
TCEAL4
Entrez Gene ID
79921
Gene Name
transcription elongation factor A like 4
Gene Aliases
NPD017, WEX7
Chromosome Location
Chr. X: 103576231 - 103587736 on Build GRCh38
UniGene ID
Hs.194329
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001006935.2 NP_001006936.1 3 720
NM_001006937.2 NP_001006938.1 3 715
NM_001300901.1 NP_001287830.1 5 1148
NM_001305840.1 NP_001292769.1 4 969
NM_001305841.1 NP_001292770.1 2 642
NM_001305842.1 NP_001292771.1 3 774
NM_024863.5 NP_079139.4 3 779
XM_017029856.1 XP_016885345.1 3 1039
XM_017029857.1 XP_016885346.1 3 1050
GenBank mRNA AF271783.1 - 3 692
AF314542.1 - 5 1148
AK024827.1 - 3 713
AK093021.1 - 1 1646
AK225117.1 - 3 713
AL833640.1 - 3 773
AY885169.1 - 4 596
BC012296.1 - 3 679 3445403
BX104938.1 - 1 167

View Full Product Details