Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001007527.1 | NP_001007528.1 | 2 | 499 | |
XM_011514162.2 | XP_011512464.1 | 2 | 941 | ||
XM_017010024.1 | XP_016865513.1 | 2 | 445 | ||
GenBank mRNA | AK091295.1 | - | 2 | 439 | |
AK307575.1 | - | 2 | 445 | ||
BC110506.2 | - | 2 | 438 | 40028737 | |
BX647340.1 | - | 2 | 474 | ||
CR749399.1 | - | 2 | 499 |