Gene Transcripts

Gene Symbol
UBE2QL1
Entrez Gene ID
134111
Gene Name
ubiquitin conjugating enzyme E2 Q family like 1
Gene Aliases
Chromosome Location
Chr. 5: 6448623 - 6496721 on Build GRCh38
UniGene ID
Hs.126856
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145161.2 NP_001138633.1 2 682
GenBank mRNA AB188494.1 - 2 1055
AK308577.1 - 2 446
BC127723.1 - 2 447
BC150554.1 - 2 448

View Full Product Details