Gene Transcripts

Gene Symbol
FAM43B
Entrez Gene ID
163933
Gene Name
family with sequence similarity 43 member B
Gene Aliases
Chromosome Location
Chr. 1: 20552439 - 20555020 on Build GRCh38
UniGene ID
Hs.429179
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207334.2 NP_997217.1 1 757
GenBank mRNA AK126900.1 - 1 757
BC142610.1 - 1 400 40147211
BC142686.1 - 1 648 40148591

View Full Product Details