Gene Transcripts

Gene Symbol
CFAP221
Entrez Gene ID
200373
Gene Name
cilia and flagella associated protein 221
Gene Aliases
FAP221, PCDP1
Chromosome Location
Chr. 2: 119544392 - 119688610 on Build GRCh38
UniGene ID
Hs.496753
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271049.1 NP_001257978.1 23 2460
NR_073132.1 24 2605
XM_006712353.3 XP_006712416.1 23 2500
XM_017003550.1 XP_016859039.1 23 2500
XM_017003551.1 XP_016859040.1 23 2627
XM_017003552.1 XP_016859041.1 23 2513
XM_017003553.1 XP_016859042.1 23 2846
XM_017003554.1 XP_016859043.1 22 2342
XM_017003555.1 XP_016859044.1 22 2342
XM_017003556.1 XP_016859045.1 22 2345
XM_017003557.1 XP_016859046.1 21 2275
XM_017003559.1 XP_016859048.1 20 3471
GenBank mRNA AK126306.1 - 10 1010
AW611757.1 - 2 384
BC036530.1 - 24 2571 4828213

View Full Product Details