Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001039960.2 | NP_001035049.1 | 25 | 3659 | |
NM_001258401.2 | NP_001245330.1 | 25 | 3605 | ||
XM_011539010.1 | XP_011537312.1 | 26 | 3714 | ||
XM_011539011.1 | XP_011537313.1 | 26 | 3659 | ||
XM_011539014.2 | XP_011537316.1 | 16 | 2312 | ||
XM_017020240.1 | XP_016875729.1 | 26 | 3741 | ||
XM_017020242.1 | XP_016875731.1 | 26 | 3728 | ||
GenBank mRNA | AK093005.1 | - | 1 | 321 | |
AK093357.1 | - | 4 | 951 | ||
AK310017.1 | - | 6 | 782 | ||
DQ063579.2 | - | 25 | 3648 | ||
DQ975204.1 | - | 25 | 3475 |