Gene Transcripts

Gene Symbol
EBLN1
Entrez Gene ID
340900
Gene Name
endogenous Bornavirus-like nucleoprotein 1
Gene Aliases
EBLN-1
Chromosome Location
Chr. 10: 22208814 - 22209983 on Build GRCh38
UniGene ID
Hs.450519
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199938.1 NP_001186867.1 1 1083

View Full Product Details