Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_032290.3 | NP_115666.2 | 2 | 111 | |
XM_017009978.1 | XP_016865467.1 | 2 | 292 | ||
XM_017009979.1 | XP_016865468.1 | 2 | 4873 | ||
XM_017009980.1 | XP_016865469.1 | 2 | 293 | ||
XM_017009981.1 | XP_016865470.1 | 2 | 143 | ||
XM_017009982.1 | XP_016865471.1 | 2 | 292 | ||
GenBank mRNA | AK297452.1 | - | 2 | 119 | |
AK299080.1 | - | 2 | 137 | ||
AL050298.1 | - | 2 | 116 | ||
BC054885.1 | - | 2 | 113 | ||
BC070332.1 | - | 2 | 149 | ||
BC107571.1 | - | 2 | 111 | ||
CR936768.1 | - | 2 | 117 | ||
DR001089.1 | - | 2 | 111 |