Gene Transcripts

Gene Symbol
ZFYVE9
Entrez Gene ID
9372
Gene Name
zinc finger FYVE-type containing 9
Gene Aliases
MADHIP, NSP, PPP1R173, SARA, SMADIP
Chromosome Location
Chr. 1: 52142001 - 52348664 on Build GRCh38
UniGene ID
Hs.532345
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004799.3 NP_004790.2 1 195
NM_007324.3 NP_015563.2 1 195
XM_017002845.1 XP_016858334.1 1 288
GenBank mRNA DR005493.1 - 1 197

View Full Product Details