Gene Transcripts

Gene Symbol
C5orf56
Entrez Gene ID
441108
Gene Name
chromosome 5 open reading frame 56
Gene Aliases
Chromosome Location
Chr. 5: 132410773 - 132476044 on Build GRCh38
UniGene ID
Hs.658288
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_045116.1 4 461
GenBank mRNA AI377899.1 - 1 339
AK128882.1 - 4 250
BM998490.1 - 1 357

View Full Product Details