Gene Transcripts

Gene Symbol
ZFAT-AS1
Entrez Gene ID
594840
Gene Name
ZFAT antisense RNA 1
Gene Aliases
NCRNA00070, SAS-ZFAT, ZFAT-AS, ZFATAS
Chromosome Location
Chr. 8: 134598071 - 134600689 on Build GRCh38
UniGene ID
Hs.626298
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002438.1 1 19
GenBank mRNA AB167742.1 - 1 19

Gene Symbol
ZFAT
Entrez Gene ID
57623
Gene Name
zinc finger and AT-hook domain containing
Gene Aliases
AITD3, ZFAT1, ZNF406
Chromosome Location
Chr. 8: 134477788 - 134713049 on Build GRCh38
UniGene ID
Hs.446172
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB167741.1 - 7 4920

View Full Product Details