Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001206559.1 | NP_001193488.1 | 1 | 196 | |
XM_005266412.1 | XP_005266469.1 | 1 | 196 | ||
XM_005266414.2 | XP_005266471.1 | 1 | 196 | ||
XM_017020623.1 | XP_016876112.1 | 1 | 152 | ||
GenBank mRNA | DR156229.1 | - | 1 | 168 |