Gene Transcripts
- Gene Symbol
- SH2B3
- Entrez Gene ID
- 10019
- Gene Name
- SH2B adaptor protein 3
- Gene Aliases
- IDDM20, LNK
- Chromosome Location
- Chr. 12: 111405106 - 111451623 on Build GRCh38
- UniGene ID
- Hs.506784
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001291424.1 |
NP_001278353.1
|
6, 7 |
850 |
|
NM_005475.2 |
NP_005466.1
|
7, 8 |
1773 |
|
XM_005253818.4 |
XP_005253875.1
|
8, 9 |
1917 |
|
XM_005253819.4 |
XP_005253876.1
|
7, 8 |
1800 |
|
XM_006719180.3 |
XP_006719243.1
|
6, 7 |
1402 |
|
XM_011537719.2 |
XP_011536021.1
|
8, 9 |
1920 |
|
XM_011537720.2 |
XP_011536022.1
|
8, 9 |
1585 |
|
XM_011537721.2 |
XP_011536023.1
|
6, 7 |
3385 |
|
GenBank mRNA |
AB208911.1 |
- |
6, 7 |
1009 |
|
AF055581.1 |
- |
7, 8 |
1773 |
|
AJ012793.1 |
- |
6, 7 |
1419 |
|
AK302173.1 |
- |
6, 7 |
850 |
|
BC136451.1 |
- |
7, 8 |
1459 |
9020440 |
View Full Product Details