Gene Transcripts

Gene Symbol
PTPMT1
Entrez Gene ID
114971
Gene Name
protein tyrosine phosphatase, mitochondrial 1
Gene Aliases
DUSP23, MOSP, PLIP, PNAS-129
Chromosome Location
Chr. 11: 47565430 - 47573461 on Build GRCh38
UniGene ID
Hs.656205
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175732.2 NP_783859.1 3, 4 634
GenBank mRNA BC014048.1 - 2, 3 280
BC020242.1 - 3, 4 463 4591438
BQ050653.1 - 1, 2 27
DA476984.1 - 3, 4 490
DR004030.1 - 3, 4 492

View Full Product Details