Gene Transcripts
- Gene Symbol
- TTC23L
- Entrez Gene ID
- 153657
- Gene Name
- tetratricopeptide repeat domain 23 like
- Gene Aliases
- MC25-1
- Chromosome Location
- Chr. 5: 34839164 - 34900511 on Build GRCh38
- UniGene ID
- Hs.435742
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001317949.1 |
NP_001304878.1
|
7, 8 |
873 |
|
NM_144725.3 |
NP_653326.3
|
7, 8 |
938 |
|
XM_011513984.2 |
XP_011512286.1
|
7, 8 |
875 |
|
XM_011513985.2 |
XP_011512287.1
|
7, 8 |
875 |
|
XM_017009119.1 |
XP_016864608.1
|
7, 8 |
1119 |
|
XM_017009120.1 |
XP_016864609.1
|
7, 8 |
938 |
|
XM_017009121.1 |
XP_016864610.1
|
7, 8 |
983 |
|
XM_017009122.1 |
XP_016864611.1
|
7, 8 |
1054 |
|
XM_017009123.1 |
XP_016864612.1
|
7, 8 |
875 |
|
XM_017009124.1 |
XP_016864613.1
|
5, 6 |
815 |
|
XM_017009125.1 |
XP_016864614.1
|
5, 6 |
616 |
|
XM_017009126.1 |
XP_016864615.1
|
5, 6 |
749 |
|
GenBank mRNA |
AK058168.1 |
- |
7, 8 |
938 |
|
BC057775.1 |
- |
7, 8 |
910 |
5295348 |
JN799997.1 |
- |
7, 8 |
873 |
|
View Full Product Details