Gene Transcripts
- Gene Symbol
- HIGD2A
- Entrez Gene ID
- 192286
- Gene Name
- HIG1 hypoxia inducible domain family member 2A
- Gene Aliases
- RCF1b
- Chromosome Location
- Chr. 5: 176388783 - 176389750 on Build GRCh38
- UniGene ID
- Hs.534575
- Species
- Homo sapiens
- Gene Symbol
- NOP16
- Entrez Gene ID
- 51491
- Gene Name
- NOP16 nucleolar protein
- Gene Aliases
- HSPC111, HSPC185
- Chromosome Location
- Chr. 5: 176383937 - 176388927 on Build GRCh38
- UniGene ID
- Hs.696283
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF151875.1 |
- |
2 |
75 |
|
View Full Product Details