Gene Transcripts
- Gene Symbol
- VWA3B
- Entrez Gene ID
- 200403
- Gene Name
- von Willebrand factor A domain containing 3B
- Gene Aliases
- SCAR22
- Chromosome Location
- Chr. 2: 98087132 - 98330675 on Build GRCh38
- UniGene ID
- Hs.269977
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_144992.4 |
NP_659429.4
|
22, 23 |
3315 |
|
XM_005263897.2 |
XP_005263954.1
|
22, 23 |
3280 |
|
XM_006712357.2 |
XP_006712420.1
|
21, 22 |
3029 |
|
XM_006712359.3 |
XP_006712422.1
|
13, 14 |
1709 |
|
XM_006712360.3 |
XP_006712423.1
|
10, 11 |
1356 |
|
XM_011510770.1 |
XP_011509072.1
|
21, 22 |
3229 |
|
XM_011510771.2 |
XP_011509073.1
|
21, 22 |
3154 |
|
XM_011510772.1 |
XP_011509074.1
|
22, 23 |
3280 |
|
XM_017003560.1 |
XP_016859049.1
|
16, 17 |
2146 |
|
XM_017003561.1 |
XP_016859050.1
|
16, 17 |
2152 |
|
XM_017003562.1 |
XP_016859051.1
|
16, 17 |
2202 |
|
XM_017003563.1 |
XP_016859052.1
|
16, 17 |
2152 |
|
GenBank mRNA |
AK090786.1 |
- |
9, 10 |
1151 |
|
AK093084.1 |
- |
10, 11 |
1282 |
|
AK098841.1 |
- |
4, 5 |
554 |
|
AK295743.1 |
- |
16, 17 |
2210 |
|
AL832635.2 |
- |
23, 24 |
3258 |
|
AL832761.1 |
- |
23, 24 |
3486 |
|
AL834173.1 |
- |
13, 14 |
1714 |
|
AL834385.1 |
- |
12, 13 |
1578 |
|
BC022028.2 |
- |
4, 5 |
545 |
|
BC150653.1 |
- |
22, 23 |
3255 |
9057024 |
View Full Product Details