Gene Transcripts

Gene Symbol
OR9Q2
Entrez Gene ID
219957
Gene Name
olfactory receptor family 9 subfamily Q member 2
Gene Aliases
OR9Q2P
Chromosome Location
Chr. 11: 58190434 - 58191518 on Build GRCh38
UniGene ID
Hs.553643
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005283.2 NP_001005283.1 1 97
GenBank mRNA BC140718.1 - 1 97

View Full Product Details