Gene Transcripts

Gene Symbol
KCNH3
Entrez Gene ID
23416
Gene Name
potassium voltage-gated channel subfamily H member 3
Gene Aliases
BEC1, ELK2, Kv12.2
Chromosome Location
Chr. 12: 49538541 - 49558312 on Build GRCh38
UniGene ID
Hs.64064
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001314030.1 NP_001300959.1 5 876
NM_012284.2 NP_036416.1 5 880
XM_011538085.2 XP_011536387.1 5 1496
XM_011538086.2 XP_011536388.1 5 1492
XM_017019096.1 XP_016874585.1 4 1262
XM_017019097.1 XP_016874586.1 5 500
GenBank mRNA AB022696.1 - 5 655
AB033108.1 - 5 880
BC150602.1 - 5 761 9056972

View Full Product Details