Gene Transcripts
- Gene Symbol
- TINF2
- Entrez Gene ID
- 26277
- Gene Name
- TERF1 interacting nuclear factor 2
- Gene Aliases
- DKCA3, TIN2
- Chromosome Location
- Chr. 14: 24239641 - 24242674 on Build GRCh38
- UniGene ID
- Hs.496191
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001099274.1 |
NP_001092744.1
|
5, 6 |
944 |
|
NM_012461.2 |
NP_036593.2
|
5, 6 |
944 |
|
XM_005267529.3 |
XP_005267586.1
|
4, 5 |
770 |
|
GenBank mRNA |
AF195512.1 |
- |
5, 6 |
864 |
|
AK023166.1 |
- |
3, 4 |
1335 |
|
AK225379.1 |
- |
5, 6 |
888 |
|
AK294119.1 |
- |
4, 5 |
770 |
|
AK315084.1 |
- |
5, 6 |
832 |
|
BC005030.1 |
- |
5, 6 |
725 |
3690254 |
BC019343.1 |
- |
5, 6 |
902 |
3505009 |
EU851975.1 |
- |
2, 3 |
118 |
|
View Full Product Details