Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000577.4 | NP_000568.1 | 5 | 572 | |
NM_001318914.1 | NP_001305843.1 | 7 | 806 | ||
NM_173841.2 | NP_776213.1 | 6 | 635 | ||
NM_173842.2 | NP_776214.1 | 4 | 564 | ||
NM_173843.2 | NP_776215.1 | 6 | 743 | ||
XM_005263661.4 | XP_005263718.1 | 5 | 591 | ||
XM_011511121.1 | XP_011509423.1 | 9 | 1094 | ||
GenBank mRNA | AF043143.1 | - | 6 | 743 | |
AK290898.1 | - | 6 | 625 | ||
AK290926.1 | - | 4 | 566 | ||
BC009745.2 | - | 5 | 477 | 3689284 | |
BI766516.1 | - | 4 | 563 | ||
M55646.1 | - | 5 | 568 | ||
X52015.1 | - | 4 | 523 | ||
X53296.1 | - | 4 | 508 | ||
X84348.1 | - | 6 | 541 |