Gene Transcripts

Gene Symbol
WEE2
Entrez Gene ID
494551
Gene Name
WEE1 homolog 2 (S. pombe)
Gene Aliases
WEE1B
Chromosome Location
Chr. 7: 141708353 - 141731271 on Build GRCh38
UniGene ID
Hs.657927
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105558.1 NP_001099028.1 6 1298
GenBank mRNA AK131218.1 - 6 1298
BM790836.1 - 3 217

View Full Product Details